Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylacetamide ≥99%, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: Acetic acid dimethylamideDimethyl Acetamide

0215092891
IC15092891EA 145.6 USD InStock
IC15092891
N,N-Dimethylacetamide ≥99%, liquid
N,N-Dimethylacetamide
Miscible with water, most organic solvents.


Formula: CH₃CON(CH₃)₂
MW: 87.12 g/mol
MDL Number: MFCD00008686
CAS Number: 127-19-5
UN: 2810
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/13/2298913.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty