Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Heptanesulfonic acid sodium salt, hydrate ~98%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: Sodium 1-heptanesulfonateSodium 1-heptanesulphonateSodium heptane-1-sulfonateSodium heptane-1-sulphonate

0215123925
IC15123925EA 192.24 USD InStock
IC15123925
1-Heptanesulfonic acid sodium salt, hydrate ~98%, white powder
1-Heptanesulphonic acid sodium salt
Sodium 1-heptanesulfonate is a commonly used ion pairing reagent for HPLC, including analyses of peptides and proteins. It has also been used in high performance capillary electrophoresis analysis of peptides.

Soluble in water (10% w/v - clear, colorless solution), ethanol or methanol (0,4% w/v - clear solution).

Caution: Research Use Only (RUO).


Formula: H₃C(CH₂)₅CH₂SO₃Na
MW: 202.25 g/mol
MDL Number: MFCD00007543
CAS Number: 22767-50-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/00/26/36420026.jpg
Order Now
Specification Test Results
Identity Test Passes
Purity ~98%
Moisture 7.4 - 9.0%
FTIR Conforms to Standard
Moisture Content 7.4-9.0%
UV/VIS:(60 mg/ml in water adjusted to pH 3) OD210 nm 1.0
OD220 nm 0.1
OD254 nm 0.04
OD280 nm 0.04

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty