Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Propionic acid ≥98.5%, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: Propanoic acid

0215195680
IC15195680EA 43.72 USD InStock
IC15195680
Propionic acid ≥98.5%, liquid
Propionic acid


Formula: C₃H₆O₂
MW: 74.08 g/mol
MDL Number: MFCD00002756
CAS Number: 79-09-4
UN: 3463
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty