Would you like to log in again?
Protocatechuate-3,4-Dioxygenase
Supplier: MP Biomedicals
0215197510 0215197505 0215197525
IC15197505EA 107.29 USD InStock
IC15197505 IC15197510 IC15197525
Protocatechuate-3,4-Dioxygenase
Enzymes
One unit will oxidize one µmole of protocatechuate to 3-carboxy-cis,cis-muconate per minute at pH 7.5 at 37 °C.
  • Inhibitors : Ag+, Hg2+, PCMB
  • Optimum temperature: 60 -65 °C
  • Thermal stability: below 50 °C (pH 6.0, 1hr)
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty