Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Reactive blue 4, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215201610
IC15201610EA 48.21 USD InStock
IC15201610
Reactive blue 4, Powder
Reactive blue 4


Formula: C₂₃H₁₄Cl₂N₆O₈S₂
MW: 637.44 g/mol
MDL Number: MFCD00001218
CAS Number: 13324-20-4
Order Now
Specification Test Results
Synonyms 1-Amino-4-[3-(4,6-dichlorotriazin-2-ylamino)-4-sulfophenylamino]anthraquinone-2-sulfonic acid; C.I. 61205; Procion® blue MX-R
Form Black Powder
Storage Store at Room Temperature (15-30 °C).
Color Index 61205
Extinction Coefficient ≥4000(Lit.)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty