Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Thiouridine ~98%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-4-thioxo-3,4-dihydropyrimidin-2(1H)-one

0215213405 0215213425 0215213480
IC15213405EA 52.12 USD InStock
IC15213405 IC15213425 IC15213480
4-Thiouridine ~98%, white powder
4-Thiouridine
4-Thio Uridine is typically used to modify oligos slated for RNA, or RNA-protein, structural studies. A 4-thio-rU modified RNA pentamer was used to study the effect of this modification on codon-anticodon interaction when it is in the wobble position of tRNA. Because 4-thio-rU is photoreactive, 4-thio-rU modified RNA oligos have also been used as photoaffinity probes in the role of substrate analogs for characterizing the enzyme:substrate complex of tRNA:pseudouridine-5S synthase . 4-thio-rU modified oligos have also been used as modules for assembling U25 small nucleolar RNAs (U25snoRNA) by ligation. These snoRNAs were used in cross-linking studies to identify which proteins assembled on them in vivo in Xenopus oocytes. In addition, because the thiol group is chemically reactive, other moieties can be conjugated at the thiol group of 4-thio-rU. Such a strategy was used to introduce spin labels to 4-thio-rU-containing RNA oligos.

Soluble in water (20 mg/ml - clear, dark yellow solution).

Caution: Research Use Only (RUO).


Formula: C₉H₁₂N₂O₅S
MW: 260.264 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00006538
CAS Number: 13957-31-8
Order Now
Specification Test Results
Identity Test Passes
Purity ~98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty