Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-thyroxine, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid

02152145.5 02152145.1 0215214501
IC15214501EA 108.12 USD InStock
IC15214501 IC152145.1 IC152145.5
L-thyroxine, Powder
L-thyroxine
Soluble in solutions of alkai hydroxides (50 mg/mL in 4M Ammonium Hydroxide in methanol); slighly insoluble in alcohol; insoluble in water, acetone, chloroform and ether.


Formula: C₁₅H₁₁I₄NO₄
MW: 776.874 g/mol
MDL Number: MFCD00002595
CAS Number: 51-48-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty