Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenol red, powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning
Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

0215255405
IC15255405EA 56.9 USD InStock
IC15255405
Phenol red, powder ACS
Phenol red
Phenol Red Sodium Salt is used as a pH indicator, it is used widely in culture media to identify changes from neutral to acidic pH values.

Phenol red in tissue culture media can act as a weak estrogen, especially with human breast cancer cells. Phenol red can be used to measure hydrogen peroxide in supernatants from cultured macrophages in multiwell plates.

It is made by condensing two moles of phenol with one mole of o-sulfobenzoic acid anhydride.

Soluble in water (3 mg/mL), ethanol (4 mg/mL), methanol or acetic acid. Readily soluble in alkaline solutions.


Formula: C₁₉H₁₄O₅S
MW: 354.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003552
CAS Number: 143-74-8
EINECS: 205-609-7
Order Now
Specification Test Results
Identity test Passes
pH Transition Intervals: Yellow 6.8 Orange 7.0 Red 8.2
Clarity of solution Passes

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty