Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride ≥99.0%, white crystalline powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: SaltHaliteSea saltRock saltSaline

0215257594 0215257591 0215257595 0215257590
IC15257591EA 102.47 USD InStock
IC15257591 IC15257590 IC15257595 IC15257594
Sodium chloride ≥99.0%, white crystalline powder ACS
Sodium chloride


Formula: NaCl
MW: 58.44 g/mol
Melting Pt: 801 °C (lit.)
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Order Now
Specification Test Results
Assay ≥99.0%
Barium Passes
Bromide ≤0.01%
Insoluble Matter ≤0.005%
Magnesium (Mg) ≤0.001%
Sulphate (SO₄) ≤0.004%
Iron (Fe) ≤0.0002%
Calcium (Ca) ≤0.002%
Chlorates and Nitrates (as NO₃) ≤0.003%
Phosphate (PO₄) 0.0005%
Heavy Metals ≤0.0005% (as Pb)
Potassium (K) ≤0.005%
Iodide (I) ≤0.002%
pH (5% aq soln, 25°C) 5.0 - 9.0

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty