Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Strontium chloride hexahydrate 99.0-103.0%, crystalline powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215258380
IC15258380EA 71.72 USD InStock
IC15258380
Strontium chloride hexahydrate 99.0-103.0%, crystalline powder ACS
Strontium chloride hexahydrate
Sulfate NMT 0.001% ;Barium NMT 0.05% ; Calcium NMT 0.05% ; Magnesium NMT0.0002%


Formula: SrCl₂·6H₂O
MW: 266.62 g/mol
MDL Number: MFCD00149865
CAS Number: 10025-70-4
Order Now
Specification Test Results
Synonyms Strontium Dichloride Hexahydrate
Form White, Hard Packed, Crystalline Powder
Storage Store at Room Temperature (15-30 °C).
Assay 99.0-103.0%
Heavy Metals NMT 0.0005% ; Iron NMT 0.0005%
Insoluble Matter NMT 0.005%
pH 5.0-7.0 (5% sol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty