Would you like to log in again?
Carmine, red powder C.I. 75470
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: Alum lake of Carminic AcidCarmine, calcium/aluminum lakeCI 75470Natural Red 4Carminic acidCochinealFD and C Carmine
0215263425
IC15263425EA 223.63 USD InStock
IC15263425
Carmine, red powder C.I. 75470
Carmine
Soluble in ethanol, alkali hydroxides, borax (0,8 mg/ ml), ammonium hydroxide (1 mg/ml), water (8 mg/ml) or ethanol (0,8 mg/ml). The addition of an acid or base can help increase the water solubility. Partly soluble in hot water; practically insoluble in cold water (8 mg/ ml) or dilute acids.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty