Would you like to log in again?
Neuraminidase
Supplier: MP Biomedicals
0215384605 0215384601
IC15384601EA 556.49 USD InStock
IC15384601 IC15384605
Neuraminidase
Enzymes
One unit will liberate 1.0 µmole of N-acetyl-neuraminic acid (NANA) per minute at pH 5.0 at 37°C, using either one NAN-lactose, bovine submaxillary mucin, or colominic acid as a substrate.
Neuraminidase enzymes are glycoside hydrolase enzymes.
Neuraminidases are used to cleave N-acetyl neuraminic acid (sialic acid) from a variety of glycoproteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty