Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetylneuraminic acid ≥98%, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: NANASialic acidNeu5AcLactaminic acid(4S,5R,6R,7S,8R)-5-Acetamido-4,6,7,8,9-pentahydroxy-2-oxononanoic acid5-Acetamido-3,5-dideoxy-D-glycero-D-galacto-nonulopyranosonic acid

0215385125
IC15385125EA 126.46 USD InStock
IC15385125
N-Acetylneuraminic acid ≥98%, Powder
N-Acetylneuraminic acid
Soluble in water


Formula: C₁₁H₁₉NO₉
MW: 309.27 g/mol
MDL Number: MFCD00006620
CAS Number: 131-48-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty