Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cinnarizine, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: 1-(Diphenylmethyl)-4-(3-phenyl-2-propenyl)piperazine1-Benzhydryl-4-cinnamylpiperazine1-Cinnamyl-4-diphenylmethylpiperazine

0215498610 0215498625
IC15498625EA 267.26 USD InStock
IC15498625 IC15498610
Cinnarizine, white powder
Cinnarizine
Ca2+ channel blocker; central and peripheral vasodilator

Soluble in chloroform (50 mg/mL).


Formula: C₂₆H₂₈N₂
MW: 368.524 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00056037
CAS Number: 298-57-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty