Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium acetate dihydrate, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: Acetic acid lithium salt

0215525683 0215525691
IC15525691EA 219.55 USD InStock
IC15525691 IC15525683
Lithium acetate dihydrate, Powder
Lithium acetate dihydrate
Water soluble 1 M, clear, colorless(Lit.)

Lithium acetate dihydrate is used in ester interchange catalyst in polyester production, anticorrosion agent in molding polyphenylene sulfide resins, and a catalyst in alkyd resin and acrylic polymer production.


Formula: H₃CCOOLi·2H₂O
MW: 102.02 g/mol
MDL Number: MFCD00066949
CAS Number: 6108-17-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty