Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyruvaldehyde (methylglyoxal), liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0215555825 0215555883
IC15555825EA 138.22 USD InStock
IC15555825 IC15555883
Pyruvaldehyde (methylglyoxal), liquid
Pyruvaldehyde (Methylglyoxal)
Methylglyoxal is the aldehyde form of pyruvic acid

Methylglyoxal is used in formation of secondary organic aerosol. It it may be involved in the formation of advanced glycation endproducts (AGEs). Pyruvic aldehyde is used in the synthesis and cytotoxicity evaluation of N-terminal analogues of tubulysin-U an antimitotic compound. Also used in the synthesis of imidazoles as potent calcitonin gene-related CGRP antagonists.


Formula: C₃H₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00006960
CAS Number: 78-98-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/99/86/36419986.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty