Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium 1-pyrrolidinecarbodithioate ≥95%, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: Pyrrolidinedithiocarbamate ammonium salt1-Pyrrolidinecarbodithioic acid, ammonium saltPyrrolidine-1-Dithiocarboxylic Acid Ammonium SaltAPCD

0215647025
IC15647025EA 407.16 USD InStock
IC15647025
Ammonium 1-pyrrolidinecarbodithioate ≥95%, white crystalline powder
Ammonium 1-pyrrolidinecarbodithioate
Pyrrolidinedithiocarbamate, ammonium salt is a complexing agent for the extraction of metal ions.

Pyrrolidinedithiocarbamate, ammonium salt is used for the extraction and pre-concentration of heavy metals with an organic solvent from aqueous solutions for atomic absorption spectroscopy.

Prevents induction of nitric oxide synthetase by inhibiting translation of NOS mRNA; induces apoptosis in rat smooth muscle cells and inhibits apoptosis in leukemia HL-60 cells.

Soluble in water (100 mg/ml with heating and vortexing - creates a tan suspension).


Formula: C₅H₁₂N₂S₂
MW: 146.3 · 18.0 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00012720
CAS Number: 5108-96-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty