Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Heptane ≥99%, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215732380
IC15732380EA 59.68 USD InStock
IC15732380
n-Heptane ≥99%, liquid
n-Heptane
Soluble in ethanol, chloroform and ether; Insoluble in water.

N - Heptane is widely applied in laboratories as a totally non-polar solvent.


Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
MDL Number: MFCD00009544
CAS Number: 142-82-5
UN: 1206
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/64/15/736415.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty