Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)], powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: EGSEthylene glycol disuccinate di(N-succinimidyl) esterBis(2,5-dioxopyrrolidin-1-yl) O,O'-ethane-1,2-diyl disuccinateDi(N-succinimidyl) ethylene glycol disuccinateEthylene glycol bis(succinimidyl succinate)

0215798283
IC15798283EA 148.13 USD InStock
IC15798283
EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)], powder
EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)]
Ethylene Glycol bis(Succinic Acid N-Hydroxysuccinimide Ester) is a homobifunctional, cleavable cross-linking reagent. It is typically, coupled to molecules containing primary amines by amide bonds buffered at pH 7.5 (6.5-8.5). The linkage provides a 12-atom spacer.

Ethylene Glycol bis(Succinic Acid N-Hydroxysuccinimide Ester) can be used to produce succinic acid 2-[3-(doxorubicin-3'-N-yl)-3-oxopropionyloxy]-ethyl ester N-hydroxysuccinimidyl ester with adriamycin at temperature of 20 °C.

Soluble in acetic acid: water (1:1): ≤50 mg/mL, DMF.

Ethylene Glycol bis(Succinic Acid N-Hydroxysuccinimide Ester) is a homobifunctional, cleavable cross-linking reagent. It is typically, coupled to molecules containing primary amines by amide bonds buffered at pH 7.5 (6.5-8.5). The linkage provides a 12-atom spacer.


Formula: C₁₈H₂₀N₂O₁₂
MW: 456.36 g/mol
MDL Number: MFCD00049057
CAS Number: 70539-42-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty