Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alitretinoin ≥98%, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: 9-cis-RAAlitretinoinNSC-659772Panretin

0215872501
IC15872501EA 87.43 USD InStock
IC15872501
Alitretinoin ≥98%, Powder
9-cis-Retinoic acid
9-cis-Retinoic acid is a high affinity ligand for the retinoid X receptor (RXR), it destabilize Vitamin D receptor RXR heterodimer-DNA complexes. It is naturally produced via isomerization of all-trans retinoic acid.

9-cis-Retinoic acid inhibits cell proliferation and induces cell differentiation.

9-cis-Retinoic acid is a high affinity ligand for the retinoid X receptor (RXR), it destabilize Vitamin D receptor RXR heterodimer-DNA complexes. It is naturally produced via isomerization of all-trans retinoic acid.

Soluble in DMSO (25 mg/ml), acetone (25 mg/ml) and ethanol (25 mg/ml). To solubilise the product in ethanol at 25 mg/ml the solution needs to be warmed to 50 °C. Store solutions aliquoted at -20 °C in the dark for up to 3 months.


Formula: C₂₀H₂₈O₂
MW: 300.442 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00270072
CAS Number: 5300-03-8
Order Now
Specification Test Results
Identity Test Passes
Purity ≥98%

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty