Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thapsigargin ≥97%, crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215899901
IC15899901EA 197.88 USD InStock
IC15899901
Thapsigargin ≥97%, crystalline powder
Thapsigargin
Thapsigargin may be used to distinguish between discrete intracellular Ca2+ pools. It induces apoptosis in rat thymocytes and in human hepatoma cells.

Potent, cell-permeable, IP3-independent intracellular calcium releaser. Blocks the transient increase in intracellular Ca2+ induced by angiostatin and endostatin. Induces apoptosis by disrupting intracellular free Ca2+ levels; incorporated into chemotherapeutic prodrug formulations.

Soluble in CH3CN (10 mg/mL-clear, colorless solution), acetonitrile (10 mg/mL - clear, colorless solution), DMSO and ethanol.

Thapsigargin may be used to distinguish between discrete intracellular Ca2+ pools. It induces apoptosis in rat thymocytes and in human hepatoma cells.


Formula: C₃₄H₅₀O₁₂
MDL Number: MFCD00083511
CAS Number: 67526-95-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/97/35/36419735.jpg
Order Now
Specification Test Results
Form White Crystalline Powder
Storage Store at -20 °C.
Refractive Index n20D 1.54 (Predicted)(Lit.)
Source Thapsia garganica

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty