Would you like to log in again?
Alkaline Phosphatase
Supplier: MP Biomedicals
0219068001
IC19068001EA 130.83 USD InStock
IC19068001
Alkaline Phosphatase
Enzymes
Dissolves readily at 5 mg/mL in a 0.025M glychine/NaOH pH 9.6 containing 1 mM magnesium chloride, 0.1 mM zinc chloride and 10% glycerol, to give a clear pale pink solution also soluble in distilled water or dilute buffer.
That amount of enzyme causing the hydrolysis of one micromole of p-nitrophenyl phosphate per minute at 25 °C and pH 9.6.
Activators: Zn, Mg2+, Ca2+, Tris buffer, diethanolamine. Inhibitors: Acidification to pH 4.5 reversibly inactives the enzyme; inorganic phosphate, chelating agents such as EDTA. Typical Contaminants: Free of endonuclease, exonuclease, and RNase activities.
Alkaline phosphatase is used to remove 5′-phosphate groups from nucleic acid molecules to prevent self-ligation of DNA fragments and vectors during recombinant DNA manipulations. It is used in the laboratory to remove phosphate monoesters which prevents self-ligation. It is also used in the dairy industry to confirm pasturization of milk. Alkaline phosphatse is commonly conjugated to antibodies and used in immunoblotting, ELISA′s, and other antibody based detection assays.
Catalyzes the cleavage of terminal phosphates from nucleic acids. The free 5′-OH can be phosphorylated with polynucleotide kinase and γ32P-ATP to produce γ32P-end-labeled nucleic acids. Linearized cloning vectors can be prevented from recircularizing by dephosphorylation with alkaline phosphatase.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty