Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Guanine, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: 2-Amino-1H-purin-6(7H)-one

0219069125
IC19069125EA 104.87 USD InStock
IC19069125
Guanine, white powder
Guanine
Guanine is one of the four main nucleobases found in the nucleic acids DNA and RNA. Guanine is a purine derivative.

Guanine is used as an additive to various products (e.g., shampoos), where it provides a pearly iridescent effect. It is also used in metallic paints and simulated pearls and plastics. It provides shimmering luster to eye shadow and nail polish.

Soluble in 0.1 N HCl (50 mg/mL - clear, colorless solution), 1 M Sodium hydroxide (50 mg/mL - clear to slightly hazy light yellow solution). Freely soluble in ammonia water, aqueous KOH solutions, dilute acids; sparingly soluble in alcohol, ether. Almost insoluble in water.


Formula: C₅H₅N₅O
MW: 151.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00071533
CAS Number: 73-40-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty