Would you like to log in again?
Cathepsin G
Supplier: MP Biomedicals
0219134480
IC19134480EA 444.97 USD InStock
IC19134480
Cathepsin G
Enzymes
The amount of enzyme that hydrolyzes one µmole of Suc-ala-ala-pro-phe-pNA (1mM) per minute 25 °C in pH 7.5, with 1.6 M NaCl.
  • Inhibitors: Chymostatin, DFP
Cathepsin G act as a bacteriocidal reagent. Catrhepsin G is often referred to as a chymotrypsin-like enzyme, since it preferentially cleaves proteins on the carboxyl side of leucine, methionine, and phenylalanine. Cathepsin G can be assayed with substrates such as azocasein, Suc-Ala-Ala-Pro-Phe-NPhNO2, and MeOSUC-Ala-Ala-Pro-Met-NPh-NO2. Cathepsin G also has a specificity of action on the oxidized B chain of insulin and on BSA, which indicates a closer similarity to porcine chymotrypsin C than to bovine chymotrypsin A.
Cathepsin G has pro-apoptotic activity and can activate caspases in vitro.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty