Would you like to log in again?
Lipoprotein-Cholesterol concentrate
Supplier: MP Biomedicals
0219147625 0219147680
IC19147680EA 1096.17 USD InStock
IC19147680 IC19147625
Lipoprotein-Cholesterol concentrate
Cell Culture Supplements and Additives
Lipoprotein-cholesterol is a mixture of lipoprotein cholesterol, phospholipids and fatty acids.
  • Filtered through 0.2 μm membrane filter
  • No preservatives
  • Free of enzyme inhibitors
  • From Bovine Serum
  • Cholesterol concentration: 1000-2000 mg/dL
For use as a serum-free or serum-reduced media nutrient supplement, calibration of cholesterol values, and enzymatic or non-enzymatic cholesterol determinations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty