Synonyms:
Cupric sulfate pentahydrate
, Copper sulfate pentahydrate
, Copper sulphate pentahydrate
, Cupric sulphate pentahydrate
0219511783
0219511790
IC19511790EA
103.47
USD
InStock
IC19511790
IC19511783
Copper(II) sulfate pentahydrate ≥99%, blue, crystalline powder
Copper (II) sulphate pentahydrate
Cupric sulfate pentahydrate is a common inorganic salt used in analytical chemistry, organic synthesis, and in art to etch zinc plates.
This compound has been used to research the copper-induced chromosomal aberrations in the fish, Channa punctata, and as a catalyst in the synthesis of ynamides and Z-enamides in the amidation of alkynyl bromides, and to catalyze the formation of ligand arm-functionalized azoamides.
Soluble in water, methanol, glycerol; slightly soluble in ethanol.