Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rifampicin, orange powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning
Synonyms: Rifamycin AMP3-(4-Methylpiperazinyliminomethyl)rifamycin SVRifampin

0219549025 02195490.2 02195490.1 0219549001
IC195490.2EA 58.41 USD InStock
IC195490.2 IC19549025 IC195490.1 IC19549001
Rifampicin, orange powder
Rifampicin
Rifampin is a bactericidal antibiotic drug of the rifamycin group. It specifically inhibits DNA-dependent bacterial RNA Polymerase. Mammalian RNA polymerase is not affected.

Rifampicin is used to treat Tuberculosis and Tuberculosis-related mycobacterial infections. It is widely used as an antipruritic agent in the autoimmune cholestatic liver disease, primary biliary cirrhosis (PBC). It has been shown to cause hepatitis.

Mode of Action: Inhibits initiation of RNA synthesis by binding to β-subunit of RNA polymerase. It Inhibits the assembly of DNA and protein into mature virus particles. EmM (max absorbance, phosphate buffer,pH 7.38): 33.20 (237 nm); 32.10 (255 nm); 27.00 (334 nm);15.40 (475 nm)(Lit.). pKa (in water):1.7 (4-hydroxyl group), 7.9 (4-piperazine nitrogen); in methylcellosolve-water (4:1): 3.6 (4-hydroxyl group), 6.7 (3-piperazine nitrogen)(Lit.).

Soluble in chloroform(349 mg/mL, 25 °C), DMSO(~100mg/mL), ethyl acetate(108 mg/mL, 25 °C), methanol(16 mg/mL, 25 °C) tetrahydrofuran.


Formula: C₄₃H₅₈N₄O₁₂
MW: 822.95 g/mol
Melting Pt: 183…188 °C (dec., lit.)
MDL Number: MFCD00151389
CAS Number: 13292-46-1
Order Now
Specification Test Results
Synonyms Rifamycin AMP; Rifampicin; 3-(4-Methylpiperazinyliminomethyl)rifamycin SV; Rifampicin
Form Orange Powder
Storage Store at +4 °C.
Isoelectric Point 4.8(in water)(Lit.)
Loss on Drying ≤2%
Optical Rotation [α]D25°=+10.6° (c=0.5% in CDCl3)(Lit.)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty