Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl green zinc chloride salt, dark powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: CI 42590Methyl green zinc chloride double saltMethyl GreenEthyl Green

04806408
IC806408EA 178.2 USD InStock
IC806408
Methyl green zinc chloride salt, dark powder
Methyl Green zinc chloride salt
Methyl green may be used as a pH indicator. Methyl green solution is a suitable counterstain for chloroacetate esterase, nonspecific esterase, alkaline phosphatase, peroxidase, naphthol AS acetate esterase, and acid phosphatase

  • Soluble in water


Formula: C₂₇H₃₅BrClN₃·xZnCl₂
MW: 653.24
MDL Number: MFCD00151094
CAS Number: 7114-03-6
Order Now
Specification Test Results
Synonyms CI 42590; 4-[[4-(Dimethylamino) phenyl][4-(dimethyliminio) cyclohexa-2,5-dien-1-ylidene] methyl]- N-ethyl-N,N- bromide chloride, compound with zinc chloride; Ethyl Green
Form Dark Powder
Storage Store at Room Temperature (15-30 °C).
Color Index 42590
Extinction Coefficient E1% = 800 - 1100 (water)Lit.
Typical Working Concentration A typical working concentration for using methyl green as a counter stain is 0.1- 0.5%.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty