Would you like to log in again?
Scoop, Stainless Steel, Bürkle
Supplier: Bürkle
5354-1000
10217-178EA 679.58 USD InStock
10217-178
Scoop, Stainless Steel, Bürkle
Scoops
The stainless steel scoop has no openings, undercuts or crevices in which residues could collect
Pore-free finishing produced by high-quality laser welding seams. The stainless steel scoop can be flame-sterilised and can therefore be made completely germ-free.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty