Would you like to log in again?
Diversified Biotech Pipetter Guide
Supplier: Diversified Biotech
GUIDE-300
490014-050EA 309.67 USD InStock
490014-050
Diversified Biotech Pipetter Guide
Pipettes Pipette Controllers
This guide enables users to easily target microplate wells using the plastic sliding grid. The sliding guide is clearly marked with letters for fast and easy orientation. Ideal for long-term pipetting.
  • Reduces eyestrain during tedious pipetting work
  • Suitable for use with multi channel pipettes
  • Adjustable height to accommodate flat or v-bottom micro or PCR plates
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty