Would you like to log in again?
Economy Cable Lockout, with 10' Cable, Brady Worldwide®
Supplier: Brady
45352
10154-262EA 55.97 USD InStock
10154-262
Economy Cable Lockout, with 10' Cable, Brady Worldwide®
Security Locks
Cable lockouts are incredibly versatile making them perfect for all of your unusual energy isolating devices that are difficult to lock out
6 lock holes in handle are 9/32" in diameter to accommodate most lock shackles and hasps.
Ordering information: Economy Cable Lockout includes 10' cable. Additional 6' and 10' cables are sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty