Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Antibody (RPE (R-Phycoerythrin))

Supplier: Columbia Biosciences
D5-1903 D5-1902 D5-1900 D5-1901
10159-220EA 531.45 USD InStock
10159-220 10159-222 10159-216 10159-218
Anti-IgG Antibody (RPE (R-Phycoerythrin))
Antibodies
Emission: 578 nm
Excitation: 565>498 nm

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host:
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty