Would you like to log in again?
Solar LED Jar Light Kit (Pre-Wired)
Supplier: SUNDANCE SOLAR MO
900-10017-35-BL
470118-650CS 336.95 USD InStock
470118-650
Solar LED Jar Light Kit (Pre-Wired)
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Teacher Developed!
  • A portion of the proceeds from the sales of this kit are being donated to help provide solar lighting solutions to disadvantaged communities in Africa.
  • This kit is pre-wired and DOES NOT require soldering.
Developed in collaboration with a science teacher from Pennsylvania, our build your own solar LED Jar light is a fun and creative way to teach and learn about solar electricity. Use your own mason jar or recycled jar and build a colorful LED light that automatically comes on at night and goes off at dawn.
Comes with everything you need. It includes: 3V 70mA Polycrystalline Solar Panel with wires, Dusk/dawn circuit board with wires and BLUE LED's, 2 AAA NiMH rechargeable batteries, Battery holder & wires, and wire nuts.
Tools required: Hot glue gun or silicone adhesive, electric drill.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty