Would you like to log in again?
Solar LED Flashlight Kit (Pre-Wired)
Supplier: SUNDANCE SOLAR MO
900-10017-40
470118-600CS 350.75 USD InStock
470118-600
Solar LED Flashlight Kit (Pre-Wired)
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Teacher Developed!
  • Kit includes everything you need (except the bottle) and comes with complete, easy to follow instructions.
  • This kit comes pre-wired and does not require soldering.
Build your own solar charged triple beam LED flashlight using a recycled drink bottle. Includes 3.6V 50mA flexible thin-film solar panel (Made in the USA) 3 bright white LED's and 2 rechargeable AAA NiMH batteries. Learn about solar energy and build a working solar flashlight!
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty