Would you like to log in again?
Stopower™ Plug Lockout, Brady Worldwide®
Supplier: Brady
65673 65673
12222-746EA 52.62 USD InStock
CA12222-746 12222-746
Stopower™ Plug Lockout, Brady Worldwide®
Security Locks
The Stopower™ Plug Lockout device is simple to use and operate
Simply slide over cord prongs and lock with key. As a result, the cord cannot be plugged in with Stopower™ in place. The device cannot be removed without a key. Standard devices are individually keyed. Multiple Stopower™ plug lockouts that are keyed alike are also available (contact your Avantor sales representative for information).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty