Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Detector H. Pylori™, Immunostics

Supplier: Immunostics
H. Pylori™
IHP-10
10147-070KT 145.62 USD InStock
10147-070
Detector H. Pylori™, Immunostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
Detector H. Pylori™ qualitatively detects anti-Helicobacter pylori antibody in human whole blood, serum, or plasma. The test is intended for use as an aid in the diagnosis of H. pylori infection in adult patients with symptoms of gastrointestinal disorders.

  • Sensitivity 95.9%
  • Specificity 89.1%
  • Test time 10 Minutes

Certifications: CLIA Waived (For Whole Blood Only). CE Marked.

Ordering information: Kit includes 10 Cassettes, 10 Disposable Pipettes, 1 Vial Developing Buffer, 1 Vial Positive Control, 1 Vial Negative Control, and 1 Product Insert.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty