Would you like to log in again?
AccuTarget™ Human siRNA Subset Library, Bioneer
Supplier: Bioneer
SHS-011053-6 SHS-011053-1 SHS-011013-6 SHS-011013-5 SHS-019013-2 SHS-011053-5 SHS-019053-2 SHS-019013-1 SHS-011053-2 SHS-011013-4 SHS-011013-3 SHS-011053-4 SHS-011013-8 SHS-011053-8 SHS-011013-1 SHS-011053-7 SHS-011053-3 SHS-019053-1 SHS-011013-7 SHS-011013-2
95030-408EA 773.88 USD InStock
95030-408 95030-490 95030-486 95030-442 95030-420 95030-410 95030-484 95030-482 95030-480 95030-492 95030-406 95030-418 95030-416 95030-512 95030-414 95030-488 95030-444 95030-510 95030-478 95030-412
AccuTarget™ Human siRNA Subset Library, Bioneer
Nucleic Acid Reagents RNAi
AccuTarget™ Human siRNA subset libraries are functional/family subsets of the Kinase or phosphatase libraries
These are available at 0.1, 0.25, 0.5 and 1 nmole guaranteed yield.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty