Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Cystine dihydrochloride 98.0-102.0%, Bioreagent

Supplier: VWR
Synonyms: (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid) dihydrochloride(2R,2'R)-3,3'-Disulfanediylbis(2-aminopropanoic acid) dihydrochloride

VWRVVWRB21502-1KG VWRVVWRB21502-5KG VWRV21502-VK
VWRB21502-1KGEA 888.12 USD InStock
VWRB21502-1KG VWRB21502-5KG VWRB21502-100G
L-Cystine dihydrochloride 98.0-102.0%, Bioreagent
L-Cystine dihydrochloride
Suitable for bioprocessing. BSE/TSE Free. Manufactured in USA. Custom Packaging and Testing Available. Change Management. Sample availability.


Formula: C₆H₁₂N₂O₄S₂·2HCl
MW: 313.23 g/mol
MDL Number: MFCD00070399
CAS Number: 30925-07-6
Order Now
Specification Test Results
Appearance white to slightly yellow crystalline
Assay (on dried basis, by titration) 98.0% to 102.0%
Identification (by IR) matches reference IR Spectrum
Solubility (100mg/L in deionized H2O) clear and complete
Loss On Drying (@105ºC/3hrs.) NMT 1.0%
Residue On Ignition NMT 0.1%
Specific Rotation (C=2, 1N HCl, 20ºC, dried @ 105ºC) 225º to -210º
Heavy Metals (as Pb) NMT 10 ppm
Bioburden NMT 100 CFU/g
Chloride (Cl) 22.2% to 23.5%
Sulfate (SO4) NMT 0.030%
Cell Culture Toxicity Tested (@ 0.06 g/L) Passes test
Endotoxin (0.1% solution in H2O, KTA) NMT 0.1 EU/mg

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty