Would you like to log in again?
ActivArmr® 27-607 Industrial gloves, oil repellent, Ansell
Supplier: Ansell
ActivArmr®
207310
32889-900CS 2006.35 USD InStock
32889-900
ActivArmr® 27-607 Industrial gloves, oil repellent, Ansell
Gloves
These nitrile gloves are three times more durable than heavy-duty leather, made extra tough for heavy-duty jobs involving rough, abrasive materials.
  • Silicone-free
  • Superior resistance to abrasion
  • More flexible than PVC
  • Excellent barrier to grease and oil
Featuring a unique Ansell nitrile formulation, gloves offer superior resistance to sharp and abrasive materials.
Use in the automotive and transportation, building and construction, machinery and equipment, and metal fabrication industries. Uses include cold plastic mouldings; construction materials, concrete, bricks and tiles; heavy-duty cables; rough castings and cores; rough timber, dry walling and plywood; steel bars, sheet metals, cans and other metal products; and street cleaning and refuse collection.
Stronger and more flexible than PVC, gloves also provide an excellent barrier to grease and oil and do not degrade like leather or cotton. Made without silicone. Guaranteed clean for metals and motor manufacture.
Certifications: ANSI level 1 cut protection; ANSI Level 4 Abrasion protection.
Packaging: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty