Would you like to log in again?
AlphaTec® 39-124 Chemical resistant gloves, Ansell
Supplier: Ansell
AlphaTec®
217806 217807
32890-166CS 1282.05 USD InStock
32890-166 32890-164
AlphaTec® 39-124 Chemical resistant gloves, Ansell
Gloves
Gloves provide heavy-duty mechanical and chemical protection for a wide variety of work environments and offer optimum comfort of use thanks to their natural shape, making gloves well-suited for use with long-lasting tasks.
  • Optimum comfort due to natural shape
  • Durable for long tasks
Superior dexterity and high levels of wet and dry grip contribute to wearer safety and ultimately to overall productivity.
Use in the agriculture and viticulture, automotive and transportation, building and construction, chemical, food processing and service, metal fabrication, and agrochemicals industries. Uses include chemical processing and preparation; degreasing; maintenance; metal fabrication; sub-assembly handling; and tank filling.
Certifications: ANSI level 1 cut protection; ANSI Level 4 Abrasion protection.
Packaging: Gloves are packaged 72 pairs in a carton.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty