Would you like to log in again?
Differential Model
Supplier: NADA SCIENTIFIC LTD.
N98-OSK16871
470157-238EA 3986.79 USD InStock
470157-238
Differential Model
Educational Materials Physics Educational Materials Mechanics Learning Activities
This product is a model of a differential. This model permits the instructor and student to manually turn the handle and apply a load to the transparent wheels to demonstrate the differential mechanism.
In automotive mechanics, the differential gear arrangement permits power from the engine to be transmitted to a pair of driving wheels, which divides the force equally between them and permits them to follow paths of different lengths, as when turning a corner or crossing an uneven road. On a straight road, the wheels rotate at the same speed. When turning a corner, the outside wheel has farther to go and will turn faster than the inner wheel if unrestrained. These items are made special to order and are not held in stock. Lead times are approximately two to three months. Freight charges may apply.
Removal of the Gear Assembly: The instructor can remove the gear assembly from the main body to highlight details and it sits comfortably on a table.
Dimensions: 28L x 22W x 25H cm
Weight: 5 kg
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty