Would you like to log in again?
Wankel Engine, Rotary Model
Supplier: NADA SCIENTIFIC LTD.
N98-OSK14943
470157-240EA 3753.29 USD InStock
470157-240
Wankel Engine, Rotary Model
Educational Materials Physics Educational Materials Mechanics Learning Activities
This product is a model of a wankel engine. The transparent plastic housing enables the students to see the internal structure at all angles. The rotor starts moving when the switch is turned on. It is powered by 110V AC. These items are made special to order and are not held in stock. Lead times are approximately two to three months. Freight charges may apply. Dimentions: Size: 15L x 20W x 28H cm ; Weight: 3 kg
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty