Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Methyltetrahydrofuran, EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger

1.08292.4000 1.08292.2500 1.08292.4000 1.08292.2500 1.08292.1000 1.08292.1000
EM1.08292.2500EA 416 USD InStock
CA1.08292.1000 EM1.08292.2500 CA1.08292.4000 EM1.08292.1000 EM1.08292.4000 CA1.08292.2500
(±)-2-Methyltetrahydrofuran, EMPLURA®, Supelco®
(±)-2-Methyltetrahydrofuran
EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.


Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 79 °C (1013 hPa)
Melting Pt: –136 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: –12 °C (closed cup)
MDL Number: MFCD00005367
CAS Number: 96-47-9
EINECS: 202-507-4
UN: 2536
ADR: 3,II
Order Now
Specification Test Results
Hill Formula C₅H₁₀O
Molar Mass 86.13 g/mol
Boiling point 78 - 80 °C
Density 0.85 g/cm³ (20 °C)
Explosion limit 1.2 - 5.7 %(V)
Flash point -12 °C
Ignition temperature 270 °C
Melting Point -136 °C
Vapor pressure 130 hPa (25 °C)
Solubility 130 g/l (25 °C)
LD 50 oral LD50 Rat300 - 2000 mg/kg
LD 50 dermal LD50 Rat2000 mg/kg
Storage class 3 Flammable liquids
WGK WGK 2 water endangering
R Phrase R 11-19-22-38-41
S Phrase S 16-29-33-26-39

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty