Would you like to log in again?
SatPax® 3000, Presaturated Wipers, Berkshire
Supplier: Berkshire
SPX300000112
21912-110CS 1067.12 USD InStock
21912-110
SatPax® 3000, Presaturated Wipers, Berkshire
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Berkshire SatPax® 3000 combines the UltraSeal® 3000 sealed-edge, cleanroom-laundered wiper (with two-ply pinsonic tubular construction) with various concentrations of isopropyl alcohol (IPA)- the SPX3000.001.12 contains a 70% IPA solution and the SPX3000.002.12 contains a 9% IPA solution.
  • Critically low particles, fibers, ions, and extractables
  • Patented sealed edge process combined with tubular design offers excellent abrasion resistance for the toughest cleaning applications
  • Pinsonic design adds surface texture that aids in stubborn residue removal
  • Reduces alcohol usage and preparation/handling costs
  • Reduces VOC emissions
  • Increases cleaning efficiency and cleaning protocol consistency
This pre-wetted format provides a more cost-effective and easy-to-use solution compared to the manual wetting of dry wipers, which requires bulk handling of messy solvents and maintenance of squirt bottles and results in inconsistent wetting and cleaning.
The cleanroom wiper is composed of 100% continuous filament polyester knit fabric.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty