Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-C9orf98 Rabbit Polyclonal Antibody

Supplier: Proteintech
21036-1-AP-150UL
10154-914EA 453 USD InStock
10154-914
Anti-C9orf98 Rabbit Polyclonal Antibody
Antibodies
The C9orf98 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human C9orf98. This antibody recognizes human antigen. The C9orf98 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 2000

Type: Primary
Antigen: C9orf98
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty