Would you like to log in again?
Scrubber, SMS, VELP Scientifica
Supplier: VELP Scientifica
F307A0660
10184-078EA 1998.45 USD InStock
10184-078
Scrubber, SMS, VELP Scientifica
Digestion Systems Mineralization Workstations
Designed to neutralize corrosive and toxic fumes produced during oxidative mineralizations or similar processes.
  • Complete system
  • Convenient to use
Neutralization with acids or bases. Optional absorption with activated carbon. Ideal for high fume levels such as with soybean analysis. Condensate flasks removable for ease of emptying. Easy to replace the neutralizing reagents. Small footprint.
Ordering information: Complete your preferred system with JP Recirculating Water Pump that aspirates fumes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty