Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AP1G2 Rabbit Polyclonal Antibody

Supplier: Proteintech
17907-1-AP-150UL
10154-778EA 453 USD InStock
10154-778
Anti-AP1G2 Rabbit Polyclonal Antibody
Antibodies
The AP1G2 Antibody from Proteintech is a rabbit polyclonal Antibody to a fusion protein of human AP1G2. This Antibody recognizes human,mouse,rat Antigen. The AP1G2 Antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 300 ; IHC 1 : 50

Type: Primary
Antigen: AP1G2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty