Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silicic acid hydrate, Extra Pure, precipitated heavy, Supelco®

Supplier: MilliporeSigma

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:


1.00656.1000 1.00656.9020 1.00656.1000 1.00656.9020
EM1.00656.9020EA 8282 USD InStock
CA1.00656.1000 CA1.00656.9020 EM1.00656.9020 EM1.00656.1000
Silicic acid hydrate, Extra Pure, precipitated heavy, Supelco®
Silicic acid hydrate


Formula: SiO₂·xH₂O
MW: 60.09 g/mol (anhy)
MDL Number: MFCD00054122
CAS Number: 10279-57-9
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 1710 °C
Molar Mass 60.08 g/mol
Bulk Density 60 kg/m3
Density 2 g/cm3 (20 °C)
Hill Formula O2Si *x H2O
Chemical Formula SiO2 * x H2O

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty