Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ytterbium(III) oxide ≥99%, Sigma-Aldrich®

Supplier: MilliporeSigma

M-Clarity™ quality level = MQ200

1.12410.0005 1.12410.0005
EM1.12410.0005CS 54800 USD InStock
CA1.12410.0005 EM1.12410.0005
Ytterbium(III) oxide ≥99%, Sigma-Aldrich®
Ytterbium (III) oxide


Formula: O₃Yb₂
MW: 394.08 g/mol
MDL Number: MFCD00011290
CAS Number: 1314-37-0
Order Now
Specification Test Results
Molar Mass 394.07 g/mol
Hill Formula O3Yb2
Chemical Formula Yb2O3

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty