Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pentadecanoic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: Pentadecylic acidn-Pentadecanoic acid

M-Clarity™ quality level = MQ200

8.00661.0010 8.00661.0010
EM8.00661.0010CS 13889 USD InStock
EM8.00661.0010 CA8.00661.0010
Pentadecanoic acid for synthesis, Sigma-Aldrich®
Pentadecanoic acid


Formula: C₁₅H₃₀O₂
MW: 242.4 g/mol
MDL Number: MFCD00002745
CAS Number: 1002-84-2
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 51 - 53 °C
Molar Mass 242.4 g/mol
Boiling Point 257 °C (133 hPa)
Flash Point 110 °C
Hill Formula C15H30O2
Chemical Formula CH3(CH2)13COOH

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty